Product Details

SNP ID
rs5749421
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:32391951 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACCTAAGTATACAATTGTGCAAAA[A/G]CAAGAGTCAGCTCAAACTACTCTTG
Phenotype
MIM: 613901
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC339666 PubMed Links

Gene Details

Gene
LOC339666
Gene Name
uncharacterized LOC339666
There are no transcripts associated with this gene.

Gene
RTCB
Gene Name
RNA 2',3'-cyclic phosphate and 5'-OH ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014306.4 Intron NP_055121.1

View Full Product Details