Product Details

SNP ID
rs3764917
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:162173826 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GAGAATATGCATTGTTTGCATGTGA[C/T]GAATGTCATTTCATTAACCAACAAG
Phenotype
MIM: 600403
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAP PubMed Links

Gene Details

Gene
FAP
Gene Name
fibroblast activation protein alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291807.1 Intron NP_001278736.1
NM_004460.3 Intron NP_004451.2
XM_011510796.2 Intron XP_011509098.1
XM_011510797.2 Intron XP_011509099.1
XM_017003585.1 Intron XP_016859074.1
Gene
LOC101929532
Gene Name
uncharacterized LOC101929532
There are no transcripts associated with this gene.

View Full Product Details