Product Details

SNP ID
rs3803907
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4250920 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCTCTTGAAGGAGGGAGCTCCCC[A/G]TTGCTGGAGGCAAGCAAGCAGTGAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC94 PubMed Links

Gene Details

Gene
CCDC94
Gene Name
coiled-coil domain containing 94
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018074.4 Intron NP_060544.2

View Full Product Details