Product Details

SNP ID
rs4773143
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:110308338 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGGTTCCCAGCGTCAATCCTGTG[C/T]TTTGCAAGCGTCGGCCTTTCACGGG
Phenotype
MIM: 120130 MIM: 120090
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COL4A1 PubMed Links

Gene Details

Gene
COL4A1
Gene Name
collagen type IV alpha 1 chain
There are no transcripts associated with this gene.

Gene
COL4A2
Gene Name
collagen type IV alpha 2
There are no transcripts associated with this gene.

View Full Product Details