Product Details

SNP ID
rs34648853
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:2833896 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCTAGGCGGGCGAGGTCGGAGTT[G/T]AGAGTGTAACTCTCTTCCAGTTTGA
Phenotype
MIM: 130135
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SERPINB1 PubMed Links

Gene Details

Gene
SERPINB1
Gene Name
serpin family B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030666.3 1232 Silent Mutation NP_109591.1
XM_011514333.1 1232 Silent Mutation XP_011512635.1
XM_011514334.2 1232 Silent Mutation XP_011512636.1
XM_011514335.2 1232 Silent Mutation XP_011512637.1

View Full Product Details