Product Details

SNP ID
rs3217744
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:127788208 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTAAAGGACCCACTCTTGCCCTTC[C/T]TGCAGATCCTGCATAGGGACATGAA
Phenotype
MIM: 603251 MIM: 613405 MIM: 604722
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDK9 PubMed Links

Gene Details

Gene
CDK9
Gene Name
cyclin dependent kinase 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261.3 Intron NP_001252.1
XM_017014184.1 Intron XP_016869673.1
Gene
MIR2861
Gene Name
microRNA 2861
There are no transcripts associated with this gene.

Gene
MIR3960
Gene Name
microRNA 3960
There are no transcripts associated with this gene.

Gene
SH2D3C
Gene Name
SH2 domain containing 3C
There are no transcripts associated with this gene.

View Full Product Details