Product Details
- SNP ID
-
hCV25592861
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:40391796 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCTTAAAACTTTGTTCTAATTTCT[C/G]AATTTTTTATCAGCAAGAACAGTTT
- Phenotype
-
MIM: 126430
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TOP2A
PubMed Links
Gene Details
- Gene
- TOP2A
- Gene Name
- topoisomerase (DNA) II alpha
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001067.3 |
|
Intron |
|
|
NP_001058.2 |
View Full Product Details