Product Details

SNP ID
rs34147280
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40392277 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGATGGGACAAAATCTTCATCATC[A/C]GTTTTTTCATCAAAATCTGAGAAAT
Phenotype
MIM: 126430
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TOP2A PubMed Links

Gene Details

Gene
TOP2A
Gene Name
topoisomerase (DNA) II alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001067.3 4130 Silent Mutation NP_001058.2
XM_005257632.1 4130 Silent Mutation XP_005257689.1
XM_011525165.1 4130 Silent Mutation XP_011523467.1

View Full Product Details