Product Details

SNP ID
hCV25593217
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:66439753 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATCGGAGAGGGGGTCTGTTCCTC[C/T]TGCTCCTCCAGCTGTTTTTCTTGGG
Phenotype
MIM: 601739
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MEIS1 PubMed Links

Gene Details

Gene
MEIS1
Gene Name
Meis homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002398.2 Intron NP_002389.1
Gene
MEIS1-AS2
Gene Name
MEIS1 antisense RNA 2
There are no transcripts associated with this gene.

Gene
MEIS1-AS3
Gene Name
MEIS1 antisense RNA 3
There are no transcripts associated with this gene.

View Full Product Details