Product Details

SNP ID
rs4451431
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136430228 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGACCCTCTTAGCTCATGGGACGA[C/T]GGCACGACCCCCAGGCCCAAGGGGG
Phenotype
MIM: 613037 MIM: 613036 MIM: 612854
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
INPP5E PubMed Links

Gene Details

Gene
INPP5E
Gene Name
inositol polyphosphate-5-phosphatase E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318502.1 Intron NP_001305431.1
NM_019892.5 Intron NP_063945.2
XM_017014926.1 Intron XP_016870415.1
Gene
PMPCA
Gene Name
peptidase, mitochondrial processing alpha subunit
There are no transcripts associated with this gene.

Gene
SEC16A
Gene Name
SEC16 homolog A, endoplasmic reticulum export factor
There are no transcripts associated with this gene.

View Full Product Details