Product Details
- SNP ID
-
rs201336440
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:137568878 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCCGGCGCTGACCGTATTTTACCC[C/G]CCTTGGGTTTTTGCCTTTTGCAGGT
- Phenotype
-
MIM: 300265
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZIC3
PubMed Links
Gene Details
- Gene
- ZIC3
- Gene Name
- Zic family member 3
View Full Product Details