Product Details

SNP ID
rs34163229
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73647154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGGGTGTTGCCCGAGGCCCCTGG[G/T]ATTGGGCCTTAGGGAGAGTTCGGGC
Phenotype
MIM: 605603
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MYOZ1 PubMed Links

Gene Details

Gene
MYOZ1
Gene Name
myozenin 1
There are no transcripts associated with this gene.

Gene
SYNPO2L
Gene Name
synaptopodin 2 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114133.2 1814 Missense Mutation TAC,TCC Y833S NP_001107605.1
NM_024875.4 1814 Missense Mutation TAC,TCC Y609S NP_079151.2
XM_005270158.2 1814 Missense Mutation TAC,TCC Y537S XP_005270215.1
XM_005270159.3 1814 Missense Mutation TAC,TCC Y537S XP_005270216.1

View Full Product Details