Product Details

SNP ID
rs112722725
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52361356 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATGGCCCCCACTGAGGAGCCAGG[A/G]GAGGGTGAAACAGTTGGGGGAACAC
Phenotype
MIM: 602767
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRT85 PubMed Links

Gene Details

Gene
KRT85
Gene Name
keratin 85
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300810.1 Intron NP_001287739.1
NM_002283.3 Intron NP_002274.1

View Full Product Details