Product Details

SNP ID
rs61733980
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30993413 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCTCGCGGATGCTGGTCTCCAG[C/T]TTGATGATCTCATTGTGCCTCGTCT
Phenotype
MIM: 607764 MIM: 611052 MIM: 601485
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HSD3B7 PubMed Links

Gene Details

Gene
HSD3B7
Gene Name
hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 7
There are no transcripts associated with this gene.

Gene
SETD1A
Gene Name
SET domain containing 1A
There are no transcripts associated with this gene.

Gene
STX1B
Gene Name
syntaxin 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052874.4 851 Silent Mutation AAA,AAG K203K NP_443106.1
XM_017022893.1 851 Silent Mutation AAA,AAG K197K XP_016878382.1

View Full Product Details