Product Details

SNP ID
rs138908675
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1578235 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGGCTCTTCTAGGGAGATGGGAA[A/G]CTCTTGGGAGGCACCCGTCATCCCA
Phenotype
MIM: 603573
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MBD3 PubMed Links

Gene Details

Gene
MBD3
Gene Name
methyl-CpG binding domain protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281453.1 Intron NP_001268382.1
NM_001281454.1 Intron NP_001268383.1

View Full Product Details