Product Details

SNP ID
rs4909953
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:10798837 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATTCATTGTGTAGCTTTACTTAAT[A/C]ATAGCTTCAAATAGTGCAGAATGAA
Phenotype
MIM: 602325
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
EIF4G2 PubMed Links

Gene Details

Gene
EIF4G2
Gene Name
eukaryotic translation initiation factor 4 gamma 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042559.2 Intron NP_001036024.3
NM_001172705.1 Intron NP_001166176.1
NM_001418.3 Intron NP_001409.3
Gene
SNORD97
Gene Name
small nucleolar RNA, C/D box 97
There are no transcripts associated with this gene.

View Full Product Details