Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001190880.2 | 11662 | Intron | NP_001177809.1 | ||
NM_001243526.1 | 11662 | Intron | NP_001230455.1 | ||
NM_031207.5 | 11662 | Intron | NP_112484.3 | ||
XM_005271239.4 | 11662 | Intron | XP_005271296.1 | ||
XM_005271240.4 | 11662 | Intron | XP_005271297.1 | ||
XM_006710937.3 | 11662 | Intron | XP_006711000.1 | ||
XM_011542222.2 | 11662 | Intron | XP_011540524.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_015284.3 | 11662 | UTR 3 | NP_056099.3 | ||
XM_005270686.3 | 11662 | Intron | XP_005270743.1 | ||
XM_006710501.3 | 11662 | Intron | XP_006710564.1 | ||
XM_011541106.2 | 11662 | Intron | XP_011539408.1 | ||
XM_011541107.2 | 11662 | Intron | XP_011539409.1 | ||
XM_017000819.1 | 11662 | Intron | XP_016856308.1 | ||
XM_017000820.1 | 11662 | Intron | XP_016856309.1 | ||
XM_017000821.1 | 11662 | Intron | XP_016856310.1 |