Product Details

SNP ID
rs16916583
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:94130735 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACTTCCTTCCACACAACTGCAGAA[C/T]GTAACTGAAAAGCAGGACAGTATTT
Phenotype
MIM: 603017
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDH17 PubMed Links

Gene Details

Gene
CDH17
Gene Name
cadherin 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144663.1 2415 Silent Mutation ACA,ACG T763T NP_001138135.1
NM_004063.3 2415 Silent Mutation ACA,ACG T763T NP_004054.3
XM_011516790.1 2415 Silent Mutation ACA,ACG T683T XP_011515092.1

View Full Product Details