Product Details

SNP ID
rs35327165
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:86654759 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGCCCTTGACTTCTCTGTGGGCT[C/T]GAGCAAGGACCATCCCAACTCAGGA
Phenotype
MIM: 606665
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OPN4 PubMed Links

Gene Details

Gene
OPN4
Gene Name
opsin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001030015.2 203 UTR 5 NP_001025186.1
NM_033282.3 203 UTR 5 NP_150598.1
XM_017016955.1 203 UTR 5 XP_016872444.1
XM_017016956.1 203 UTR 5 XP_016872445.1
XM_017016957.1 203 Intron XP_016872446.1

View Full Product Details