Product Details

SNP ID
rs2254953
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:62943308 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATTTCAATGCATGTGAGGGGGAG[A/G]GGTTTGAGTTTGCTCTTTTATGTAG
Phenotype
MIM: 604140 MIM: 611625 MIM: 612107
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DIDO1 PubMed Links

Gene Details

Gene
DIDO1
Gene Name
death inducer-obliterator 1
There are no transcripts associated with this gene.

Gene
GID8
Gene Name
GID complex subunit 8 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017896.2 Intron NP_060366.1
Gene
SLC17A9
Gene Name
solute carrier family 17 member 9
There are no transcripts associated with this gene.

View Full Product Details