Product Details
- SNP ID
-
rs34594201
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:94284103 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGCAGTAATTTCTGACAAGATTCC[C/T]GCCACTGCCTCAGTGCCCCTTTCAT
- Phenotype
-
MIM: 616790
MIM: 605271
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PPP4R4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117152910] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PPP4R4
- Gene Name
- protein phosphatase 4 regulatory subunit 4
There are no transcripts associated with this gene.
- Gene
- SERPINA10
- Gene Name
- serpin family A member 10
View Full Product Details