Product Details

SNP ID
rs112864059
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:65869696 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGGGCCACTCCCGGTGGACCCTC[A/G]TGCCGGCCACCCCTGCACTGATATA
Phenotype
MIM: 605570
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAB11A PubMed Links

Gene Details

Gene
RAB11A
Gene Name
RAB11A, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206836.1 Intron NP_001193765.1
NM_004663.4 Intron NP_004654.1

View Full Product Details