Product Details
- SNP ID
-
rs75216429
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
20
- Location
-
Chr.1:18634236 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAACAAAACTGGAGTCAGGGAGTGT[A/G]CTCAGTGTCTGCTCTCCATCCTCAC
- Phenotype
-
MIM: 167410
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PAX7
PubMed Links
Gene Details
- Gene
- PAX7
- Gene Name
- paired box 7
View Full Product Details