Product Details

SNP ID
rs114192056
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:22327570 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGTTCTTGCCATCTGAATTCATA[A/G]TTTTCTATTTTAATTATTTTTCAGG
Phenotype
MIM: 164831 MIM: 616700
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BMI1 PubMed Links

Gene Details

Gene
BMI1
Gene Name
BMI1 proto-oncogene, polycomb ring finger
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005180.8 Intron NP_005171.4
Gene
COMMD3
Gene Name
COMM domain containing 3
There are no transcripts associated with this gene.

Gene
COMMD3-BMI1
Gene Name
COMMD3-BMI1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204062.1 Intron NP_001190991.1

View Full Product Details