Product Details

SNP ID
rs3816993
Assay Type
Functionally tested
NCBI dbSNP Submissions
34
Location
Chr.1:219969329 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
GAATGTTTTTCCACTATCCAAAACC[C/T]ACGCTCACAGCACATCAAGACACAA
Phenotype
MIM: 138295 MIM: 611146
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EPRS PubMed Links

Gene Details

Gene
EPRS
Gene Name
glutamyl-prolyl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004446.2 Intron NP_004437.2
XM_017000614.1 Intron XP_016856103.1
Gene
SLC30A10
Gene Name
solute carrier family 30 member 10
There are no transcripts associated with this gene.

View Full Product Details