Product Details

SNP ID
rs3740204
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:11463176 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTGGCTCAGCGTGCTTTCTCGGGCT[A/G]GGAGGGTAAAGGGCATGCCGGGGGC
Phenotype
MIM: 605405
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
USP6NL PubMed Links

Gene Details

Gene
USP6NL
Gene Name
USP6 N-terminal like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080491.3 5178 Silent Mutation CCC,CCT P601P NP_001073960.1
NM_014688.3 5178 Silent Mutation CCC,CCT P584P NP_055503.1
XM_006717542.3 5178 Silent Mutation CCC,CCT P612P XP_006717605.1
XM_011519762.2 5178 Silent Mutation CCC,CCT P612P XP_011518064.1
XM_011519763.2 5178 Silent Mutation CCC,CCT P428P XP_011518065.1
XM_017016971.1 5178 Silent Mutation CCC,CCT P602P XP_016872460.1
XM_017016972.1 5178 Silent Mutation CCC,CCT P428P XP_016872461.1
XM_017016973.1 5178 Silent Mutation CCC,CCT P607P XP_016872462.1

View Full Product Details