Product Details

SNP ID
rs11570257
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:59301745 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGAGTAAGTCAGCCTTTTGTGGTGA[A/G]GAACGTGGCTCCCGGACCAGGCCCA
Phenotype
MIM: 131242
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EDN3 PubMed Links

Gene Details

Gene
EDN3
Gene Name
endothelin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302455.1 Intron NP_001289384.1
NM_001302456.1 Intron NP_001289385.1
NM_207032.2 Intron NP_996915.1
NM_207033.2 Intron NP_996916.1
NM_207034.2 Intron NP_996917.1
XM_005260312.4 Intron XP_005260369.1
XM_005260313.4 Intron XP_005260370.1
XM_006723734.3 Intron XP_006723797.1
XM_011528655.2 Intron XP_011526957.1

View Full Product Details