Product Details

SNP ID
rs35544417
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:151814201 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTGTACCTTCTGATTGTGGGGAT[A/C]CTCATAGCATATTATATTTATACGC
Phenotype
MIM: 600338
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AADAC PubMed Links

Gene Details

Gene
AADAC
Gene Name
arylacetamide deacetylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001086.2 129 Silent Mutation ATA,ATC I13I NP_001077.2
XM_005247104.4 129 Silent Mutation ATA,ATC I13I XP_005247161.1
Gene
AADACL2-AS1
Gene Name
AADACL2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details