Product Details
- SNP ID
-
hCV25608067
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:33742128 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTTGATAAGAAGAGCATCTATCA[C/G]TTTTTTAGTTATTTCATGTAAATTC
- Phenotype
-
MIM: 107271
MIM: 609089
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CD59
PubMed Links
Gene Details
- Gene
- CD59
- Gene Name
- CD59 molecule
There are no transcripts associated with this gene.
- Gene
- FBXO3
- Gene Name
- F-box protein 3
View Full Product Details