Product Details

SNP ID
rs77167399
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:103914981 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGTTCATACCTAGTTAGAACCCA[C/T]AGTTCCTTTACAATGTGAATTGTGA
Phenotype
MIM: 604573
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C14orf2 PubMed Links

Gene Details

Gene
C14orf2
Gene Name
chromosome 14 open reading frame 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127393.1 Intron NP_001120865.1
NM_004894.2 Intron NP_004885.1

View Full Product Details