Product Details

SNP ID
rs61730328
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:29673328 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGGAAGCTCTTCCCACACACAGA[A/G]CATGAATGGGGCCGGTAACCCAGAT
Phenotype
MIM: 159465 MIM: 612192
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MOG PubMed Links

Gene Details

Gene
MOG
Gene Name
myelin oligodendrocyte glycoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008228.2 749 Intron NP_001008229.1
NM_001008229.2 749 Intron NP_001008230.1
NM_001170418.1 749 Intron NP_001163889.1
NM_002433.4 749 Intron NP_002424.3
NM_206809.3 749 Intron NP_996532.2
NM_206810.3 749 Intron NP_996533.2
NM_206811.3 749 Intron NP_996534.2
NM_206812.3 749 Intron NP_996535.2
NM_206814.5 749 Intron NP_996537.3
XM_005249131.3 749 Intron XP_005249188.1
Gene
ZFP57
Gene Name
ZFP57 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001109809.2 749 Silent Mutation NP_001103279.2
XM_006715087.3 749 Silent Mutation XP_006715150.1
XM_011514570.2 749 Silent Mutation XP_011512872.1

View Full Product Details