Product Details
- SNP ID
-
rs569012373
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:66212244 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAGAAACAATCATTTCTTAAATGT[A/G]GTAAATAGCTAAGCCAAACCAAATA
- Phenotype
-
MIM: 138700
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
APOH
PubMed Links
Gene Details
- Gene
- APOH
- Gene Name
- apolipoprotein H
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000042.2 |
|
Intron |
|
|
NP_000033.2 |
View Full Product Details