Product Details

SNP ID
rs4985526
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:70388328 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTAGAAGACTCTGCCCAAGATGGTC[C/T]TGTCACCCATATTCTACCCAGGCCA
Phenotype
MIM: 607188
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ST3GAL2 PubMed Links

Gene Details

Gene
ST3GAL2
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006927.3 Intron NP_008858.1

View Full Product Details