Product Details

SNP ID
rs148089913
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:134256654 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCAGAGTGGGACCAGAAGGTAGG[C/T]GTTTTTCTTGTGCTTAGACGTTCTA
Phenotype
MIM: 604773 MIM: 613739 MIM: 610027
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ACAD8 PubMed Links

Gene Details

Gene
ACAD8
Gene Name
acyl-CoA dehydrogenase family member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014384.2 Intron NP_055199.1
XM_005271505.3 Intron XP_005271562.1
XM_011542750.2 Intron XP_011541052.1
XM_017017542.1 Intron XP_016873031.1
XM_017017543.1 Intron XP_016873032.1
XM_017017544.1 Intron XP_016873033.1
XM_017017545.1 Intron XP_016873034.1
XM_017017546.1 Intron XP_016873035.1
XM_017017547.1 Intron XP_016873036.1
XM_017017548.1 Intron XP_016873037.1
XM_017017549.1 Intron XP_016873038.1
Gene
THYN1
Gene Name
thymocyte nuclear protein 1
There are no transcripts associated with this gene.

Gene
VPS26B
Gene Name
VPS26, retromer complex component B
There are no transcripts associated with this gene.

View Full Product Details