Product Details

SNP ID
rs62620249
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:85541777 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGTAGAGGCTGTTTTAATCTCT[C/T]CTAACATTAAATTCTGGAGCTTGAT
Phenotype
MIM: 137167 MIM: 601468 MIM: 616350
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GGCX PubMed Links

Gene Details

Gene
GGCX
Gene Name
gamma-glutamyl carboxylase
There are no transcripts associated with this gene.

Gene
MAT2A
Gene Name
methionine adenosyltransferase 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005911.5 Intron NP_005902.1
Gene
PARTICL
Gene Name
promoter of MAT2A antisense radiation-induced circulating long non-coding RNA
There are no transcripts associated with this gene.

View Full Product Details