Product Details

SNP ID
rs61733131
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:54781524 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTCCACTGCGCCAGCAGCGCCT[A/C]GAGCAGGCAGCCGAAGCCGTCGGTG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TTC22 PubMed Links

Gene Details

Gene
TTC22
Gene Name
tetratricopeptide repeat domain 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114108.1 1534 Missense Mutation NP_001107580.1
NM_017904.3 1534 Intron NP_060374.2
XM_011541671.2 1534 Nonsense Mutation XP_011539973.1
XM_017001582.1 1534 Nonsense Mutation XP_016857071.1

View Full Product Details