Product Details

SNP ID
rs7972225
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:26123825 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTGGCTGTCCATTCAGCACAGCA[A/G]TTTAAGCAAACACTTTGAAAGAAGT
Phenotype
MIM: 606200 MIM: 601599
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BHLHE41 PubMed Links

Gene Details

Gene
BHLHE41
Gene Name
basic helix-loop-helix family member e41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030762.2 Intron NP_110389.1
Gene
SSPN
Gene Name
sarcospan
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135823.1 Intron NP_001129295.1
NM_005086.4 Intron NP_005077.2
XM_011520853.2 Intron XP_011519155.1
XM_011520854.2 Intron XP_011519156.1
XM_011520855.2 Intron XP_011519157.1

View Full Product Details