Product Details

SNP ID
rs61729992
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:117179291 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCGCTCTGGGCTTGCCCTTCTTG[A/G]TGCTCTTGGTGGCCTCGGTCGAGAG
Phenotype
MIM: 602508
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PAFAH1B2 PubMed Links

Gene Details

Gene
PAFAH1B2
Gene Name
platelet activating factor acetylhydrolase 1b catalytic subunit 2
There are no transcripts associated with this gene.

Gene
SIDT2
Gene Name
SID1 transmembrane family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040455.1 69 Missense Mutation ATG,GTG M10V NP_001035545.1

View Full Product Details