Product Details

SNP ID
hCV25618984
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:150402310 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCATTTGTTGTCCCTGCCCCTTT[A/C]TAACATCCTGGACCTGCAGAGCAGT
Phenotype
MIM: 142790 MIM: 606847
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CD74 PubMed Links

Gene Details

Gene
CD74
Gene Name
CD74 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025158.2 Intron NP_001020329.1
NM_001025159.2 Intron NP_001020330.1
NM_004355.3 Intron NP_004346.1
Gene
TCOF1
Gene Name
treacle ribosome biogenesis factor 1
There are no transcripts associated with this gene.

View Full Product Details