Product Details

SNP ID
rs28381070
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:113905972 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAGATAGTTAGCCATAAGTTGAG[A/G]AGATATTTTAATCCAGCCAAGGAAA
Phenotype
MIM: 607245 MIM: 609683
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AP4B1 PubMed Links

Gene Details

Gene
AP4B1
Gene Name
adaptor related protein complex 4 beta 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253852.2 Intron NP_001240781.1
NM_001253853.2 Intron NP_001240782.1
NM_001308312.1 Intron NP_001295241.1
NM_006594.4 Intron NP_006585.2
XM_011540523.2 Intron XP_011538825.1
XM_011540524.2 Intron XP_011538826.1
XM_011540525.2 Intron XP_011538827.1
XM_011540528.2 Intron XP_011538830.1
XM_017000088.1 Intron XP_016855577.1
XM_017000089.1 Intron XP_016855578.1
XM_017000090.1 Intron XP_016855579.1
XM_017000091.1 Intron XP_016855580.1
XM_017000092.1 Intron XP_016855581.1
XM_017000093.1 Intron XP_016855582.1
Gene
AP4B1-AS1
Gene Name
AP4B1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
DCLRE1B
Gene Name
DNA cross-link repair 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319946.1 Intron NP_001306875.1
NM_001319947.1 Intron NP_001306876.1
NM_022836.3 Intron NP_073747.1

View Full Product Details