Product Details
- SNP ID
-
rs17348421
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:45552174 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GCAGAAACCTTTTTTGACTTTATGG[A/G]GTAAAAGAAAACACTTGACCTTGAA
- Phenotype
-
MIM: 609031
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EPPIN
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs17433181] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EPPIN
- Gene Name
- epididymal peptidase inhibitor
There are no transcripts associated with this gene.
- Gene
- EPPIN-WFDC6
- Gene Name
- EPPIN-WFDC6 readthrough
There are no transcripts associated with this gene.
- Gene
- WFDC8
- Gene Name
- WAP four-disulfide core domain 8
View Full Product Details