Product Details

SNP ID
rs72698020
Assay Type
Functionally tested
NCBI dbSNP Submissions
26
Location
Chr.1:114569976 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTAAACAATTTAAAAGATGATGG[A/T]ATTATAGATACTCACTTTGACTCCA
Phenotype
MIM: 605783
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
BCAS2 PubMed Links

Gene Details

Gene
BCAS2
Gene Name
breast carcinoma amplified sequence 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005872.2 Intron NP_005863.1

View Full Product Details