Product Details

SNP ID
rs34205880
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3110296 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGCAGGAGGCACATTCGGGGCCC[A/G]GGATGCTCCCAGGCTGCTCTGGTAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UBOX5 PubMed Links

Gene Details

Gene
UBOX5
Gene Name
U-box domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267584.1 1460 Silent Mutation CCC,CCT P477P NP_001254513.1
NM_014948.3 1460 Missense Mutation CCG,CTG P479L NP_055763.1
NM_199415.2 1460 Missense Mutation CCG,CTG P425L NP_955447.1
Gene
UBOX5-AS1
Gene Name
UBOX5 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details