Product Details

SNP ID
rs113667176
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143822511 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCAGAGAACACAGGCCTCTCTC[A/G]GGCCACACAAGCCCTGAAGAGGGCT
Phenotype
MIM: 615563 MIM: 604819 MIM: 607733
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR937 PubMed Links

Gene Details

Gene
MIR937
Gene Name
microRNA 937
There are no transcripts associated with this gene.

Gene
NRBP2
Gene Name
nuclear receptor binding protein 2
There are no transcripts associated with this gene.

Gene
PUF60
Gene Name
poly(U) binding splicing factor 60
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136033.2 Intron NP_001129505.1
NM_001271096.1 Intron NP_001258025.1
NM_001271097.1 Intron NP_001258026.1
NM_001271098.1 Intron NP_001258027.1
NM_001271099.1 Intron NP_001258028.1
NM_001271100.1 Intron NP_001258029.1
NM_014281.4 Intron NP_055096.2
NM_078480.2 Intron NP_510965.1
XM_011516929.2 Intron XP_011515231.1
XM_011516930.2 Intron XP_011515232.1
XM_017013234.1 Intron XP_016868723.1
XM_017013235.1 Intron XP_016868724.1
XM_017013236.1 Intron XP_016868725.1
XM_017013237.1 Intron XP_016868726.1
XM_017013238.1 Intron XP_016868727.1
XM_017013239.1 Intron XP_016868728.1
XM_017013240.1 Intron XP_016868729.1
Gene
SCRIB
Gene Name
scribbled planar cell polarity protein
There are no transcripts associated with this gene.

View Full Product Details