Product Details

SNP ID
rs35647906
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:185298076 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTATAGGGGCTCCATTCATTTG[A/G]CTCAAGGTTATAGACTTCCACCGTA
Phenotype
MIM: 609209
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IVNS1ABP PubMed Links

Gene Details

Gene
IVNS1ABP
Gene Name
influenza virus NS1A binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006469.4 2376 Missense Mutation CCA,TCA P630S NP_006460.2
XM_005244843.3 2376 Missense Mutation CCA,TCA P412S XP_005244900.1
XM_011509079.2 2376 Missense Mutation CCA,TCA P637S XP_011507381.1
XM_011509080.2 2376 Missense Mutation CCA,TCA P548S XP_011507382.1
XM_017000084.1 2376 Missense Mutation CCA,TCA P419S XP_016855573.1
Gene
SWT1
Gene Name
SWT1, RNA endoribonuclease homolog
There are no transcripts associated with this gene.

View Full Product Details