Product Details

SNP ID
rs7908553
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:118008964 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTTCCTTCTTTCTTTCTCTCTCT[C/T]TGTCCAATTATCAATACAATACAAT
Phenotype
MIM: 608599
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAB11FIP2 PubMed Links

Gene Details

Gene
RAB11FIP2
Gene Name
RAB11 family interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014904.2 2013 UTR 3 NP_055719.1
XM_005269629.3 2013 UTR 3 XP_005269686.1
XM_017015926.1 2013 Intron XP_016871415.1

View Full Product Details