Product Details

SNP ID
rs17881206
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20469429 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAACTGTGCGAACCAGACCCGGCA[A/G]CCTTGCTCAGTTCAGCATAGCGGAG
Phenotype
MIM: 164050 MIM: 609865
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PNP PubMed Links

Gene Details

Gene
PNP
Gene Name
purine nucleoside phosphorylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000270.3 51 UTR 5 NP_000261.2
Gene
TMEM55B
Gene Name
transmembrane protein 55B
There are no transcripts associated with this gene.

View Full Product Details