Product Details

SNP ID
rs566743405
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:64388278 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGCGCATGCGCCACCGCGTCTGC[A/G]GGGGGAGAAGCGGGCAGGGGCGCAG
Phenotype
MIM: 172460
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MTHFD1 PubMed Links

Gene Details

Gene
MTHFD1
Gene Name
methylenetetrahydrofolate dehydrogenase, cyclohydrolase and formyltetrahydrofolate synthetase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005956.3 238 UTR 5 NP_005947.3

View Full Product Details