Product Details

SNP ID
rs34019986
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:23308109 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTGCTGAGAGTGTCAACAAGGA[A/G]ATGGAACCACTGGTGGGACAAGTGC
Phenotype
MIM: 601931
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BCL2L2 PubMed Links

Gene Details

Gene
BCL2L2
Gene Name
BCL2 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199839.1 571 Silent Mutation GAA,GAG E114E NP_001186768.1
NM_004050.4 571 Silent Mutation GAA,GAG E114E NP_004041.1
Gene
BCL2L2-PABPN1
Gene Name
BCL2L2-PABPN1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199864.1 571 Silent Mutation GAA,GAG E114E NP_001186793.1
Gene
PPP1R3E
Gene Name
protein phosphatase 1 regulatory subunit 3E
There are no transcripts associated with this gene.

View Full Product Details