Product Details

SNP ID
rs79604667
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141629468 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAACTGGGGGCTCCAGCCTAGAG[C/G]CTAGAGGCAGGGACAGGAGAGGGAT
Phenotype
MIM: 605166 MIM: 611213
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FCHSD1 PubMed Links

Gene Details

Gene
FCHSD1
Gene Name
FCH and double SH3 domains 1
There are no transcripts associated with this gene.

Gene
HDAC3
Gene Name
histone deacetylase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003883.3 Intron NP_003874.2
XM_011537697.2 Intron XP_011535999.1
Gene
RELL2
Gene Name
RELT like 2
There are no transcripts associated with this gene.

View Full Product Details